Learn More
Hampton Research HR2-944-66/Index #66 (F06), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502239
Encompass
HR2-944-66/Index #66 (F06), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-223-9 Supplier Hampton Research Supplier No. HR294466
HR2-944-66/Index #66 (F06), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.