Learn More
Hampton Research HR2-944-68/Index #68 (F08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502241
Encompass
HR2-944-68/Index #68 (F08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-1 Supplier Hampton Research Supplier No. HR294468
HR2-944-68/Index #68 (F08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.