Learn More
Hampton Research HR2-944-69/Index #69 (F09), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502242
Encompass
HR2-944-69/Index #69 (F09), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-2 Supplier Hampton Research Supplier No. HR294469
HR2-944-69/Index #69 (F09), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.