Learn More
Hampton Research HR2-944-70/Index #70 (F10), 185 ml - 0.2 M Sodium chloride, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502243
Encompass
HR2-944-70/Index #70 (F10), 185 ml - 0.2 M Sodium chloride, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-3 Supplier Hampton Research Supplier No. HR294470
HR2-944-70/Index #70 (F10), 185 ml - 0.2 M Sodium chloride, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.