Learn More
Hampton Research HR2-944-72/Index #72 (F12), 185 ml - 0.2 M Sodium chloride, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502245
Encompass
HR2-944-72/Index #72 (F12), 185 ml - 0.2 M Sodium chloride, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-5 Supplier Hampton Research Supplier No. HR294472
HR2-944-72/Index #72 (F12), 185 ml - 0.2 M Sodium chloride, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.