Learn More
Hampton Research HR2-944-74/Index #74 (G02), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502247
Encompass
HR2-944-74/Index #74 (G02), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-7 Supplier Hampton Research Supplier No. HR294474
HR2-944-74/Index #74 (G02), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 5.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.