Learn More
Hampton Research HR2-944-75/Index #75 (G03), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502248
Encompass
HR2-944-75/Index #75 (G03), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-8 Supplier Hampton Research Supplier No. HR294475
HR2-944-75/Index #75 (G03), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.