Learn More
Hampton Research HR2-944-76/Index #76 (G04), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502249
Encompass
HR2-944-76/Index #76 (G04), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-224-9 Supplier Hampton Research Supplier No. HR294476
HR2-944-76/Index #76 (G04), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.