Learn More
Hampton Research HR2-944-77/Index #77 (G05), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502250
Encompass
HR2-944-77/Index #77 (G05), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-225-0 Supplier Hampton Research Supplier No. HR294477
HR2-944-77/Index #77 (G05), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.