Learn More
Hampton Research HR2-944-79/Index #79 (G07), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502252
Encompass
HR2-944-79/Index #79 (G07), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-225-2 Supplier Hampton Research Supplier No. HR294479
HR2-944-79/Index #79 (G07), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.