Learn More
Hampton Research HR2-944-80/Index #80 (G08), 185 ml - 0.2 M Ammonium acetate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502253
Encompass
HR2-944-80/Index #80 (G08), 185 ml - 0.2 M Ammonium acetate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-225-3 Supplier Hampton Research Supplier No. HR294480
HR2-944-80/Index #80 (G08), 185 ml - 0.2 M Ammonium acetate, 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.