Learn More
Hampton Research HR2-944-81/Index #81 (G09), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502254
Encompass
HR2-944-81/Index #81 (G09), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-225-4 Supplier Hampton Research Supplier No. HR294481
HR2-944-81/Index #81 (G09), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.