Learn More
Hampton Research HR2-944-86/Index #86 (H02), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502259
Encompass
HR2-944-86/Index #86 (H02), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-225-9 Supplier Hampton Research Supplier No. HR294486
HR2-944-86/Index #86 (H02), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.