Learn More
Hampton Research HR2-944-88/Index #88 (H04), 185 ml - 0.2 M Ammonium citrate tribasic pH 7.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502261
Encompass
HR2-944-88/Index #88 (H04), 185 ml - 0.2 M Ammonium citrate tribasic pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-226-1 Supplier Hampton Research Supplier No. HR294488
HR2-944-88/Index #88 (H04), 185 ml - 0.2 M Ammonium citrate tribasic pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.