Learn More
Hampton Research HR2-944-89/Index #89 (H05), 185 ml - 0.1 M Succinic acid pH 7.0, 15% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502262
Encompass
HR2-944-89/Index #89 (H05), 185 ml - 0.1 M Succinic acid pH 7.0, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-226-2 Supplier Hampton Research Supplier No. HR294489
HR2-944-89/Index #89 (H05), 185 ml - 0.1 M Succinic acid pH 7.0, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.