Learn More
Hampton Research HR2-944-90/Index #90 (H06), 185 ml - 0.2 M Sodium formate, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502263
Encompass
HR2-944-90/Index #90 (H06), 185 ml - 0.2 M Sodium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-226-3 Supplier Hampton Research Supplier No. HR294490
HR2-944-90/Index #90 (H06), 185 ml - 0.2 M Sodium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.