Learn More
Newcomer Supply Index Tab Cards for Slides, 500/box
SDP
Catalog No. NC1814510
Encompass
Index Tab Cards allow for easy categorizing and partitioning of slides in the storage file drawers. Constructed of a heavy green cover stock, the slide Index card is designed to help the user quickly locate a specific slide within a tray. Designed to be used in the Economy (cardboard) storage drawers. May not fit properly in all metal storage drawers. Dimensions of the card: 3 1/4"(h) x 1"(w).
Catalog No. NC1814510 Supplier Newcomer Supply Supplier No. 6879
Index Tab Cards allow for easy categorizing and partitioning of slides in the storage file drawers. Constructed of a heavy green cover stock, the slide Index card is designed to help the user quickly locate a specific slide within a tray. Designed to be used in the Economy (cardboard) storage drawers. May not fit properly in all metal storage drawers. Dimensions of the card: 3 1/4"(h) x 1"(w).