Learn More
Grainger Indexable Toolholder SCBC HSS
Supplier:  Grainger 16R834
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R834 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a maximum depth of cut of 0.630 inches and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel with a black oxide finish and has an overall length of 5 inches. It can be used for facing, turning, profiling and grooving operations on lathes and CNC turning centers. The toolholder requires a screw-clamp style insert with a .312 inch thickness for proper seating.
Catalog No. 50-276-7435