Learn More
Grainger Indexable Toolholder SCBC HSS
Supplier:  Grainger 16R835
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R835 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a 35 degree lead angle and accepts negative rake inserts with a .312 inch maximum depth of cut. The toolholder body is made from steel and has a black oxide finish. It is designed for light to medium duty turning applications. The toolholder requires a screw-on style clamp to hold the insert in place.
Catalog No. 50-276-6608