Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502764901
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R844 is a right-hand turning toolholder with a 16mm shank diameter and a 0.750" insert size. It has a 0.750" IC insert pocket and accepts CCGT, CCMT, and CCGT inserts. The toolholder is made from steel with a black oxide finish and has a maximum depth of cut of 0.315". It is suitable for light to medium duty turning applications. The toolholder accepts inserts with screw clamping and has an overall length of 4". It can be used for facing, profiling, and turning operations.
Catalog No. 50-276-4901 Supplier Grainger Supplier No. 16R844
The Micro 100 Indexable Turning Toolholder 16R844 is a right-hand turning toolholder with a 16mm shank diameter and a 0.750" insert size. It has a 0.750" IC insert pocket and accepts CCGT, CCMT, and CCGT inserts. The toolholder is made from steel with a black oxide finish and has a maximum depth of cut of 0.315". It is suitable for light to medium duty turning applications. The toolholder accepts inserts with screw clamping and has an overall length of 4". It can be used for facing, profiling, and turning operations.