Learn More
Grainger Indexable Toolholder SCLC HSS
Supplier:  Grainger 16R840
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R840 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a 40 degree lead angle and accepts CNMG style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for external turning operations and accepts inserts with a .312 inch thickness. The toolholder has a maximum depth of cut of .630 inches.
Catalog No. 50-276-6935