Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502766936
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R842 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a clamping screw for securing inserts and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and allows high speed machining up to 5000 RPM. It can be used for facing, turning, profiling and grooving operations on lathes and CNC machines. The toolholder has an overall length of 4 inches and weighs 0.44 lbs. It is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6936 Supplier Grainger Supplier No. 16R842
The Micro 100 Indexable Turning Toolholder 16R842 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a clamping screw for securing inserts and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and allows high speed machining up to 5000 RPM. It can be used for facing, turning, profiling and grooving operations on lathes and CNC machines. The toolholder has an overall length of 4 inches and weighs 0.44 lbs. It is manufactured in the USA by Micro 100 Tool Corp.