Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502766937
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R845 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a positive rake angle of 7 degrees and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and has an overall length of 5". It can be used for facing, turning, profiling, and grooving applications in lathes when paired with a suitable carbide insert. The toolholder allows for precision machining with its rigid clamping design and precise insert pocket.
Catalog No. 50-276-6937 Supplier Grainger Supplier No. 16R845
The Micro 100 Indexable Turning Toolholder 16R845 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a positive rake angle of 7 degrees and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and has an overall length of 5". It can be used for facing, turning, profiling, and grooving applications in lathes when paired with a suitable carbide insert. The toolholder allows for precision machining with its rigid clamping design and precise insert pocket.