Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502766880
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R850 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a positive rake angle of 5 degrees and accepts CCGT and CCMT style inserts. The toolholder body is made from steel and has an overall length of 5". It can be used for facing, turning, profiling, and grooving applications. The toolholder allows high feed rates and depth of cuts. It is designed for use on CNC lathes and screw machines.
Catalog No. 50-276-6880 Supplier Grainger Supplier No. 16R850
The Micro 100 Indexable Turning Toolholder 16R850 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a positive rake angle of 5 degrees and accepts CCGT and CCMT style inserts. The toolholder body is made from steel and has an overall length of 5". It can be used for facing, turning, profiling, and grooving applications. The toolholder allows high feed rates and depth of cuts. It is designed for use on CNC lathes and screw machines.