Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502766015
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R839 is a right-hand turning toolholder with a 1" shank diameter and accepts 16R turning inserts. It has a maximum depth of cut of 0.500" and a maximum insert size of 0.625" x 0.625". The toolholder is made of steel and has a black oxide finish. It is suitable for general purpose turning applications. The toolholder requires a right-hand insert and has a screw clamping system to hold the insert in place. It allows high feed rates and depth of cuts. The toolholder can also be used for facing, profiling, grooving, and cutoff operations.
Catalog No. 50-276-6015 Supplier Grainger Supplier No. 16R839
The Micro 100 Indexable Turning Toolholder 16R839 is a right-hand turning toolholder with a 1" shank diameter and accepts 16R turning inserts. It has a maximum depth of cut of 0.500" and a maximum insert size of 0.625" x 0.625". The toolholder is made of steel and has a black oxide finish. It is suitable for general purpose turning applications. The toolholder requires a right-hand insert and has a screw clamping system to hold the insert in place. It allows high feed rates and depth of cuts. The toolholder can also be used for facing, profiling, grooving, and cutoff operations.