Learn More
Grainger Indexable Toolholder SCLC HSS
Supplier:  Grainger 16R846
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R846 is a right-hand turning toolholder with a 16mm shank diameter and a 0.787" insert size. It has a 0.787" x 0.787" square insert pocket and accepts CNMG and DNMG style inserts. The toolholder is made from forged steel and has a maximum depth of cut of 0.630". It is designed for general purpose turning applications. The toolholder body length is 5" and the overall length including shank is 6". It requires a minimum bore diameter of 1.260".
Catalog No. 50-276-6016