Learn More
Grainger Indexable Toolholder SCLC HSS
Catalog No. 502764899
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R841 is a right-hand turning toolholder with a 1" shank diameter and a 1/2" IC insert size. It has a maximum depth of cut of 0.500" and a minimum bore diameter of 1.250". The toolholder body is made from steel with a black oxide finish and it accepts CCGT, CCMT, DCGT, and DCMT style inserts. This is a versatile toolholder suitable for a wide range of turning operations.
Catalog No. 50-276-4899 Supplier Grainger Supplier No. 16R841
The Micro 100 Indexable Turning Toolholder 16R841 is a right-hand turning toolholder with a 1" shank diameter and a 1/2" IC insert size. It has a maximum depth of cut of 0.500" and a minimum bore diameter of 1.250". The toolholder body is made from steel with a black oxide finish and it accepts CCGT, CCMT, DCGT, and DCMT style inserts. This is a versatile toolholder suitable for a wide range of turning operations.