Learn More
Grainger Indexable Toolholder SDJC HSS
Supplier:  Grainger 16R830
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R837 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a 95 degree entering angle and accepts CCGT, CCMT, DCGT, and DCMT style inserts. The toolholder body is made from steel and has a black oxide finish. It is suitable for light to medium duty turning applications. The toolholder allows for precision insert indexing and efficient chip removal. Its secure clamping system ensures accurate insert positioning. Overall, this is a versatile indexable turning toolholder for use on lathes when paired with the appropriate insert.
Catalog No. 50-276-6625