Learn More
Grainger Indexable Toolholder SDJC HSS
Catalog No. 502766797
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R838 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a 38mm max depth of cut and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for external turning operations and accepts inserts with screw clamping. The toolholder allows high feed rates and depth of cuts. It is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6797 Supplier Grainger Supplier No. 16R832
The Micro 100 Indexable Turning Toolholder 16R838 is a right-hand turning toolholder with a 16mm shank diameter and 8mm insert size. It has a 38mm max depth of cut and accepts CCGT, CCMT, and TPMT style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for external turning operations and accepts inserts with screw clamping. The toolholder allows high feed rates and depth of cuts. It is manufactured in the USA by Micro 100 Tool Corp.