Learn More
Grainger Indexable Toolholder SDJC HSS
Catalog No. 502766919
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R829 is a right-hand turning toolholder with a 1" shank diameter and 1.25" maximum cutting depth. It has a 0.625" IC insert size and accepts CCGT, CCMT, and CCWT style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for general purpose turning applications. The toolholder allows high feed rates and depth of cuts. It has a maximum insert size of 0.625" x 0.625" and a minimum bore diameter of 1". The toolholder can be used for facing, profiling, grooving, parting off, and turning.
Catalog No. 50-276-6919 Supplier Grainger Supplier No. 16R829
The Micro 100 Indexable Turning Toolholder 16R829 is a right-hand turning toolholder with a 1" shank diameter and 1.25" maximum cutting depth. It has a 0.625" IC insert size and accepts CCGT, CCMT, and CCWT style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for general purpose turning applications. The toolholder allows high feed rates and depth of cuts. It has a maximum insert size of 0.625" x 0.625" and a minimum bore diameter of 1". The toolholder can be used for facing, profiling, grooving, parting off, and turning.