Learn More
Grainger Indexable Toolholder SDJC HSS
Supplier:  Grainger 16R831
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R831 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a 0.625" maximum depth of cut and can hold negative rake inserts. The toolholder body is made from steel and has a black oxide finish. It accepts CCGT, CCMT, DCGT, and DCMT style inserts for versatile turning applications. The toolholder allows high feed rates and depth of cuts. It is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6948