Learn More
Grainger Indexable Toolholder SDNC HSS
Catalog No. 502765643
Encompass_Preferred
The Grainger website features indexable turning profiling toolholders for CNC lathes. These toolholders have a precision ground body and use indexable inserts for the cutting edges. They are offered in a variety of styles including positive rake, neutral rake, and negative rake designs. The toolholders come in multiple sizes to fit different lathe turret stations. They are made from durable materials like steel and carbide and allow for quick insert changes. The profiling toolholders provide versatile turning, facing, profiling, and grooving operations. Overall, these are high-quality toolholders designed for robust and accurate CNC machining applications.
Catalog No. 50-276-5643 Supplier Grainger Supplier No. 16T406
The Grainger website features indexable turning profiling toolholders for CNC lathes. These toolholders have a precision ground body and use indexable inserts for the cutting edges. They are offered in a variety of styles including positive rake, neutral rake, and negative rake designs. The toolholders come in multiple sizes to fit different lathe turret stations. They are made from durable materials like steel and carbide and allow for quick insert changes. The profiling toolholders provide versatile turning, facing, profiling, and grooving operations. Overall, these are high-quality toolholders designed for robust and accurate CNC machining applications.