Learn More
StatLab India Ink Controls
SDP
Catalog No. 22130488
Change view
Click to view available options
Species:
Candida albicans
Cryptococcus species
Cryptococcus species, Candida albicans
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
22-130-488 Cryptococcus species
22-130-512 Candida albicans
22-130-513 Cryptococcus species, Candida albicans
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-488 Supplier StatLab Supplier No. 756
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
India Ink can be used to detect microorganisms in various body fluids.
  • India Ink Controls
Choose between positive control, negative control, or set of both:
  • Positive control is Cryptococcus sp. to visualize capsule production and includes 1 mL bottle
  • Negative Control is C. albicans and includes 1 mL bottle
  • Set contains 1 mL bottle of negative control and 1 mL bottle of positive control
Specifications
Product Type Positive Control
Sterility Non-sterile
Includes 1 mL Positive Control
Species Cryptococcus species