Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals India Ink soln., 0.2% in PBS buffer

Catalog No. AAJ61007K2
Change view
Click to view available options
Quantity:
500mL
1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61007K2 1L
AAJ61007AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61007K2 Supplier Thermo Scientific Chemicals Supplier No. J61007K2
  • $259.50
    $196.65 / Each of 1

    Save $62.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

India ink is a simple black ink. Previously it has been used for writing and printing and is now commonly used for drawing in comic books and comic strips.

India ink is a simple black ink. Previously it has been used for writing and printing and is now commonly used for drawing in comic books and comic strips. India ink is also used in medical research to visualize blood vessels when viewed under a microscope. It is used in a diluted form as an ultra-fine polishing medium for making precise optical surfaces on metals. In ophthalmology, it is used to some extent in corneal tattooing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1L
Format Liquid
Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.