Learn More
Sigma Aldrich Fine Chemicals Biosciences Indican Assay Kit sufficie
Catalog No. 502904185
Encompass
Indican (indoxyl sulfate) is a putrefaction product resulting from bacterial deconjugation of tryptophan in the small intestine. Indican is water soluble and excreted through urine. Increases in urine indican can indicate protein maldigestion or malabsortion or the overgrowth of gut anaerobic bacteria. Indican levels in urine can also increase in response to a high protein diet.
Catalog No. 50-290-4185 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1281KT
Indican (indoxyl sulfate) is a putrefaction product resulting from bacterial deconjugation of tryptophan in the small intestine. Indican is water soluble and excreted through urine. Increases in urine indican can indicate protein maldigestion or malabsortion or the overgrowth of gut anaerobic bacteria. Indican levels in urine can also increase in response to a high protein diet.