Learn More
Sklar Instruments Sklar Dry Heat indicator strips are used with with dry heat processes. BX100.
SDP
Supplier:  Sklar Instruments 101117
Encompass_Preferred
Sklars Dry Heat indicator strips are used with with dry heat processes. Strips change from green to brown when processed. These strips are 4 inches long.
Catalog No. 50-109-4462