Learn More
Mopec Indicating Desiccant Packets
SDP
Catalog No. 22446299 Shop All Mopec Products
Change view
Click to view available options
Quantity:
1000/Cs.
Catalog No. Quantity
22-446-299 1000/Cs.
1 options
Catalog No. 22-446-299 Supplier Mopec Supplier No. FS404
Add to Cart
Add to Cart
Provides moisture protection while allowing visual monitoring of absorption capacity. Color-changing indicator beads signal when the moisture-absorbing material has reached capacity, making it easier to identify when replacement is needed while helping protect stored items from humidity.
Construction
  • Each packet contains 1 gram of cobaltous silica gel with millions of pores
  • Adheres to FDA guidelines
Quantity
  • 1,000 per case
Dimensions
  • 1.75 L x 0.75 W x 0.25 H in.
Performance
  • Absorbs water vapor
  • Eliminates moisture and dampness
  • Gel turns pink when moisture is absorbed
Specifications
Material Silica Gel
Quantity 1000/Cs.
Grade Ungraded