Learn More
Gilson Company Inc Indicating Drierite, No. 4 Granules, 5 lb
Catalog No. NC9061953
Encompass
Indicating Drierite is used to rapidly absorb water vapor from air and gases. Made from cobalt chloride impregnated granules, the blue Drierite crystals change to a distinct pink color when exhausted. Indicating Drierite can be regenerated in an oven for longer life or can be replaced as needed.
Catalog No. NC9061953 Supplier Gilson Company Inc Supplier No. SGA105
Indicating Drierite is used to rapidly absorb water vapor from air and gases. Made from cobalt chloride impregnated granules, the blue Drierite crystals change to a distinct pink color when exhausted. Indicating Drierite can be regenerated in an oven for longer life or can be replaced as needed.