Learn More
Med Vet International McKesson Sterilization Biological Indicator Vial, 73-SCS025, 250/Case
Catalog No. 503044254
Encompass_Preferred
McKesson Sterilization Biological Indicator Vial Steam Product features include final read-out in only 24 hours and fits existing incubators Vials monitor steam sterilization processes 25 vials per box
Catalog No. 50-304-4254 Supplier Med Vet International Supplier No. 524877CS
 Restricted Item
Add to Cart
Add to Cart
McKesson Sterilization Biological Indicator Vial Steam Product features include final read-out in only 24 hours and fits existing incubators Vials monitor steam sterilization processes 25 vials per box