Learn More
Med Vet International McKesson Sterilization Biological Indicator Vial, 73-SCS100, 400/Case
Catalog No. 503045058
Encompass_Preferred
McKesson Sterilization Biological Indicator Vial Steam Product features include final read-out in only 24 hours and fits existing incubators Vials monitor steam sterilization processes 100 vials per box
Catalog No. 50-304-5058 Supplier Med Vet International Supplier No. 524878CS
 Restricted Item
Add to Cart
Add to Cart
McKesson Sterilization Biological Indicator Vial Steam Product features include final read-out in only 24 hours and fits existing incubators Vials monitor steam sterilization processes 100 vials per box