Learn More
Med Vet International Attest Super Rapid Readout Sterilization Biological Indicator Vial Steam, 1492V, 200/Case
Catalog No. 502832265
Encompass_Preferred
The self-contained design includes a carrier with spores of Geobacillus stearothermophilus and a media ampoule containing bacteriological media Provides biological indicator results in 24 minutes in 3M Attest 490/490H auto-readers with software version 4.2.7 or greater Dynamic-air-removal (pre-vacuum and SFPP): 132? C (270? F) 3 or 4 minutes 135? C (275? F) 3 minutes Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive Puts your instrument turnaround speed into overdrive for improved patient safety
Catalog No. 50-283-2265 Supplier Med Vet International Supplier No. 852219CS
The self-contained design includes a carrier with spores of Geobacillus stearothermophilus and a media ampoule containing bacteriological media Provides biological indicator results in 24 minutes in 3M Attest 490/490H auto-readers with software version 4.2.7 or greater Dynamic-air-removal (pre-vacuum and SFPP): 132? C (270? F) 3 or 4 minutes 135? C (275? F) 3 minutes Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive Puts your instrument turnaround speed into overdrive for improved patient safety