Learn More
Med Vet International Attest Rapid Readout Sterilization Biological Indicator Vial Vaporized Hydrogen Peroxide, 1295, 120/Case
Catalog No. 502831149
Encompass_Preferred
The Attest rapid readout biological indicator has several spores that enable them to provide quick results Its faster biological indicator results allow you to standardize release practices This biological indicator allows you to document results on the same day so it simplifies workflow The connection to instrument tracking systems helps avoid documentation errors Its auto-reader detects the presence of the enzyme
Catalog No. 50-283-1149 Supplier Med Vet International Supplier No. 1029909CS
Add to Cart
Add to Cart
Request a Quote
The Attest rapid readout biological indicator has several spores that enable them to provide quick results Its faster biological indicator results allow you to standardize release practices This biological indicator allows you to document results on the same day so it simplifies workflow The connection to instrument tracking systems helps avoid documentation errors Its auto-reader detects the presence of the enzyme