Learn More
Med Vet International Attest Super Rapid 5 Steam-Plus Sterilization Indicator Challenge Pack Steam 2 inch, 41482V, 24/Case
Catalog No. 502832266
Encompass_Preferred
Each challenge pack contains a 3M™ Attest™ Super Rapid Readout Biological Indicator 1492V (brown cap), a 3M™ Comply™ SteriGage™ Steam Chemical Integrator, and a record keeping sheet Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality
Catalog No. 50-283-2266 Supplier Med Vet International Supplier No. 852279CS
Each challenge pack contains a 3M™ Attest™ Super Rapid Readout Biological Indicator 1492V (brown cap), a 3M™ Comply™ SteriGage™ Steam Chemical Integrator, and a record keeping sheet Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality