Learn More
Med Vet International Attest Super Rapid 5 Steam-Plus Sterilization Indicator Challenge Pack Steam 2 inch, 41482Vf, 24/Case
Catalog No. 502831183
Encompass_Preferred
Each challenge pack contains a 3M™ Attest™ Super Rapid Readout Biological Indicator (1492V), a 3M™ Comply™ SteriGage™ Steam Chemical Integrator, and a record keeping sheet Specifically designed for routinely challenging and conducting qualification testing of 270°F (132°C) and 275°F (135°C) dynamic air removal (prevacuum) steam sterilization processes in healthcare facilities Provides BI results in 24 minutes in 3M Attest 490/490H Auto-readers with software version 4.2.7 or greater Dynamic-air-removal (Pre-vacuum and SFPP): 132C (270F) 4 min. 135C (275F) 3 min Contains a 3M™ Attest ™ Super Rapid Readout Biological Indicator 1492V and a Type 5 chemical integrator The biological indicator within the challenge pack contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive
Catalog No. 50-283-1183 Supplier Med Vet International Supplier No. 1026077CS
 Restricted Item
Add to Cart
Add to Cart
Each challenge pack contains a 3M™ Attest™ Super Rapid Readout Biological Indicator (1492V), a 3M™ Comply™ SteriGage™ Steam Chemical Integrator, and a record keeping sheet Specifically designed for routinely challenging and conducting qualification testing of 270°F (132°C) and 275°F (135°C) dynamic air removal (prevacuum) steam sterilization processes in healthcare facilities Provides BI results in 24 minutes in 3M Attest 490/490H Auto-readers with software version 4.2.7 or greater Dynamic-air-removal (Pre-vacuum and SFPP): 132C (270F) 4 min. 135C (275F) 3 min Contains a 3M™ Attest ™ Super Rapid Readout Biological Indicator 1492V and a Type 5 chemical integrator The biological indicator within the challenge pack contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive