Learn More
Med Vet International Attest Sterilization Biological Indicator Vial Steam, 1261, 400/Case
Catalog No. 502831336
Encompass_Preferred
Blue cap Self-contained indicators significantly reduce possibility of contamination, minimizing false positive and assuring more accurate results
Catalog No. 50-283-1336 Supplier Med Vet International Supplier No. 149569CS
Blue cap Self-contained indicators significantly reduce possibility of contamination, minimizing false positive and assuring more accurate results