Learn More
Med Vet International McKesson SCBI Incubator Kit, 73-SK115, Each
Catalog No. 503044467
Encompass_Preferred
SCBI Incubator Kit McKesson Dry block incubator has a light to indicate when it's ready to use and a vial-crushing slot that allows prepare BI vial There are 25 BI vials for use in monitoring steam sterilization processes ¾-inch steam tape will let you know if the optimum temperature was reached in your autoclave Steam sterilization integrator is for placement inside packs prior to processing; indicator moves into the teal safe zone This kit has everything you need to monitor steam sterilization processes, including a notebook and an educational CD-ROM
Catalog No. 50-304-4467 Supplier Med Vet International Supplier No. 541269EA
 Restricted Item
Add to Cart
Add to Cart
SCBI Incubator Kit McKesson Dry block incubator has a light to indicate when it's ready to use and a vial-crushing slot that allows prepare BI vial There are 25 BI vials for use in monitoring steam sterilization processes ¾-inch steam tape will let you know if the optimum temperature was reached in your autoclave Steam sterilization integrator is for placement inside packs prior to processing; indicator moves into the teal safe zone This kit has everything you need to monitor steam sterilization processes, including a notebook and an educational CD-ROM