Learn More
Med Vet International Attest Sterilization Biological Indicator Vial Steam, 1262, 100/Box
Catalog No. 502830844
Encompass_Preferred
The Attest™ Biological Indicator has an auto-reader that helps detect the presence of the enzyme Its spore burden provides an extra margin of safety This biological indicator has a definitive color change to help indicate the sterilization process The optimized media helps promote rapid growth of spores Its record-keeping sheet helps standardize release practices and simplify workflows The connection to instrument tracking systems helps avoid documentation errors
Catalog No. 50-283-0844 Supplier Med Vet International Supplier No. 134484BX
 Restricted Item
Add to Cart
Add to Cart
The Attest™ Biological Indicator has an auto-reader that helps detect the presence of the enzyme Its spore burden provides an extra margin of safety This biological indicator has a definitive color change to help indicate the sterilization process The optimized media helps promote rapid growth of spores Its record-keeping sheet helps standardize release practices and simplify workflows The connection to instrument tracking systems helps avoid documentation errors