Learn More
Med Vet International Attest Sterilization Biological Indicator Vial Steam, 1261P, 100/Case
Catalog No. 502829502
Encompass_Preferred
Blue cap Biological indicator vials are easy to use and interpret with visual color readout in 24 hours Self-contained biological indicators significantly reduce possibility of contamination, minimizing false positives and assuring more accurate results Monitors 270°F/132° C gravity steam sterilization processes Use in conjunction with 3M™ Attest™ Biological Indicator Incubator 116
Catalog No. 50-282-9502 Supplier Med Vet International Supplier No. 153736CS
Blue cap Biological indicator vials are easy to use and interpret with visual color readout in 24 hours Self-contained biological indicators significantly reduce possibility of contamination, minimizing false positives and assuring more accurate results Monitors 270°F/132° C gravity steam sterilization processes Use in conjunction with 3M™ Attest™ Biological Indicator Incubator 116