Learn More
Med Vet International Attest Rapid Readout Sterilization Biological Indicator Vial Steam 2-1/2 inch, 1491, 200/Case
Catalog No. 502831376
Encompass_Preferred
Provides biological indicator results in 24 minutes in 3M Attest 490/490H Auto-readers with software version 4.2.7 or greater Helps deliver biological indicator results often before a patient leaves the surgical suite- Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive
Catalog No. 50-283-1376 Supplier Med Vet International Supplier No. 1009962CS
Provides biological indicator results in 24 minutes in 3M Attest 490/490H Auto-readers with software version 4.2.7 or greater Helps deliver biological indicator results often before a patient leaves the surgical suite- Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality Puts your instrument turnaround speed into overdrive