Learn More
MSC JT&M Indicator Positioner, Cast Iron Base, Hardened Post, 1 ea.
Catalog No. 5000512868
Encompass
This indicator positioner and holder provides a stable platform for precision measurement applications. Designed for durability, the unit features a robust construction suitable for industrial dimensional measurement tasks.
  • Constructed with a durable cast iron base and hardened post
  • Offers a height adjustment range of 5/8 in. to 5 in.
  • Compatible with 3/8 in. diameter indicators
  • Manufactured in the USA
Catalog No. 50-005-12868 Supplier MSC Supplier No. 05868260
This indicator positioner and holder provides a stable platform for precision measurement applications. Designed for durability, the unit features a robust construction suitable for industrial dimensional measurement tasks.
  • Constructed with a durable cast iron base and hardened post
  • Offers a height adjustment range of 5/8 in. to 5 in.
  • Compatible with 3/8 in. diameter indicators
  • Manufactured in the USA